Learn More
Carl Zeiss™ Objective A Achromat
Use these universal objectives for routine research. Zeiss™ A-Plan Objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm.
2928.94 SEK
Product Code | Brand | Magnification Power | Description | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Description | Type | Price | Quantity & Availability | |||
11805063
|
Carl Zeiss™
421041-9910-000 |
10X | Numerical Aperture: 0.25 mm | Ph1 M27 |
2928.94 SEK
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
Description
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- Allow contrasting methods ranging from brightfield to phase contrast and for excitation wavelength within the visible spectrum
- Can be used for epifluorescence work
Specifications
1 Ea. |