missing translation for 'onlineSavingsMsg'
Läs mer

Oct-4B Antibody (OTI7H1), Allophycocyanin, Novus Biologicals™

Produktkod. 30028684 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
Förpackningsstorlek:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30028684 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30028684 Supplier Novus Biologicals Supplier No. NBP273141APC

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Oct-4B Monoclonal antibody specifically detects Oct-4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-4B
Användningsområden Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona OTI7H1
Konjugera APC
Utspädning Western Blot, Immunocytochemistry/ Immunofluorescence
Formulering PBS
Gene Alias OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1
Värd art Mouse
Immunogen Full length human recombinant protein of human POU5F1(NP_002692) produced in HEK293T cell.
Reningsmetod Immunogen affinity purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 5460
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4C in the dark.
Form Purified
Isotyp IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.