Learn More
OCT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540642
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: YLPRGPGGGA GGTGPLMHPD AAAAAAAAAE RLHAGAAYRE VQKLMHHEWL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
OCT6 Polyclonal antibody specifically detects OCT6 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
OCT6 | |
Polyclonal | |
Unconjugated | |
POU3F1 | |
6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
Rabbit | |
Protein A | |
RUO | |
5453 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q03052 | |
POU3F1 | |
Synthetic peptide directed towards the N-terminal of human POU3F1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |