missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | OCT6 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838814
|
Proteintech
18997-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828814
|
Proteintech
18997-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to OCT6.
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.Specifications
OCT6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P20267, P21952, Q03052 | |
18991, 192110, 5453 | |
Peptide | |
Primary | |
-20°C | |
POU3F1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Oct 6, Oct6, Octamer binding protein 6, OTF 6, OTF6, POU class 3 homeobox 1, POU3F1, SCIP | |
Pou3f1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |