Läs mer
Octachlorocyclopentene(>90%), TRC
Produktkod. 30417534
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417534 500 mg
30420518 100 mg
30417978 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO233615500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 706-78-5
Molekylformel C5Cl8
Molekylvikt (g/mol) 343.67
Synonym Cyclopentene, 1,2,3,3,4,4,5,5-octachloro- (ACI), 1,2,3,3,4,4,5,5-Octachlorocyclopentene (ACI), Cyclopentene, octachloro- (6CI, 8CI, 9CI), NSC 4736, Octachlorocyclopentene, Perchlorocyclopentene
IUPAC-namn 1,2,3,3,4,4,5,5-octachlorocyclopentene
LEDER ClC1=C(Cl)C(Cl)(Cl)C(Cl)(Cl)C1(Cl)Cl
Specifikationer
Kemiskt namn eller material Octachlorocyclopentene
Kvantitet 500 mg
InChI formel InChI=1S/C5Cl8/c6-1-2(7)4(10,11)5(12,13)3(1,8)9
Formel vikt 339.7508