missing translation for 'onlineSavingsMsg'
Läs mer

Octachloronapthalene, TRC

Produktkod. 30410783
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410783 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30410783 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO2336005G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Octachloronaphthalene
CAS 2234-13-1
Anteckningar om renhetsgrad HPLC
Molekylformel C10 Cl8
Kvantitet 5 g
Synonym Naphthalene','1,2,3,4,5,6,7,8-octachloro-, Naphthalene','octachloro- (6CI,7CI,8CI,9CI), 1,2,3,4,5,6,7,8-Octachloronaphthalene, Halowax 1051, NSC 243655, Octachloronaphthalene, PCN 75, Perchloronaphthalene, Perna, PCN No. 75
InChI formel InChI=1S/C10Cl8/c11-3-1-2(5(13)9(17)7(3)15)6(14)10(18)8(16)4(1)12
LEDER Clc1c(Cl)c(Cl)c2c(Cl)c(Cl)c(Cl)c(Cl)c2c1Cl
IUPAC-namn 1,2,3,4,5,6,7,8-octachloronaphthalene
Molekylvikt (g/mol) 403.73
Formel vikt 399.7508
Procent renhet >95
Visa mer Visa mindre
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.