missing translation for 'onlineSavingsMsg'
Läs mer

Oktadekametylnonasiloxan, TRC

Product Code. 30136677
Change view
Click to view available options
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30136677 500 mg
30006396 100 mg
30007017 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136677 Supplier Toronto Research Chemicals Supplier No. TRCO401575500MG
 Begränsad artikel
Beräknad leverans: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-71-8
Molekylformel C18H54O9Si9
Molekylvikt (g/mol) 667.39
Synonym 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16,18,18-Octadecamethylcyclononasiloxane (ACI), Cyclononasiloxane, octadecamethyl- (8CI, 9CI), Octadecamethylcyclononasiloxane, Octamethylcyclononasiloxane
IUPAC-namn 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16,18,18-oktakametyl-1,3,5,7,9,11,13,15,17-nonaoxa-2,4,6,8,10,12,14,16,18-nonasilacyklooktakane
LEDER C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Kemiskt namn eller material Octadecamethylnonasiloxane
Kvantitet 500 mg
InChI formel InChI=1S/C18H54O9Si9/c1-28(2)19-29(3,4)21-31(7,8)23-33(11,12)25-35(15,16)27-36(17,18)26-34(13,14)24-32(9,10)22-30(5,6)20-28/h1-18H3
Formel vikt 666.1691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.