Läs mer
Octadecanedioic Acid Mono-L-carnitine Ester Chloride, TRC
Produktkod. 30419770
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30419770 25 mg
1 options
Produktkod. 30419770 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23526025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Octadecanedioic Acid Mono-L-carnitine Ester Chloride
CAS 123233-50-1
Molekylformel C25H48ClNO6
Kvantitet 25 mg
Synonym (R)-3-Carboxy-2-[(17-carboxy-1-oxoheptadecyl)oxy]-N,N,N-trimethyl-1-propanaminium Chloride, (2R)-3-Carboxy-2-[(17-carboxy-1-oxoheptadecyl)oxy]-N,N,N-trimethyl-1-propanaminium Chloride, (R)-3-((17-Carboxyheptadecanoyl)oxy)-4-(trimethylammonio)butanoate Chloride
InChI formel InChI=1S/C25H47NO6.ClH/c1-26(2,3)21-22(20-24(29)30)32-25(31)19-17-15-13-11-9-7-5-4-6-8-10-12-14-16-18-23(27)28;/h22H,4-21H2,1-3H3,(H-,27,28,29,30);1H/t22-;/m1./s1
LEDER O=C(CCCCCCCCCCCCCCCCC(O)=O)O[C@H](CC(O)=O)C[N+](C)(C)C.[Cl-]
IUPAC-namn (R)-3-carboxy-2-((17-carboxyheptadecanoyl)oxy)-N,N,N-trimethylpropan-1-aminium chloride
Molekylvikt (g/mol) 494.1
Formel vikt 493.92