Läs mer
Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, TRC
Produktkod. 30135979
Change view
Kvantitet:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135979 50 g
17978664 5 g
17977814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135979 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23969550G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2082-79-3
Molekylformel C35 H62 O3
Molekylvikt (g/mol) 530.865
Synonym Plastic additive 11, 3,5-bis(1,1-Dimethylethyl)-4-hydroxybenzenepropanoic acid octadecyl ester, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic acid octadecyl ester, 2,6-Di-tert-butyl-4-(octadecanoxycarbonylethyl)phenol, 2,6-Di-tert-butyl-4-[[(octadecyloxy)carbonyl]ethyl]phenol, 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid octadecyl ester, 3-(3,5-Di-tert-butyl-4-hydroxyphenol)propionic acid octadecyl ester, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid octadecyl ester, 4-Carboxyethyl)-2,6-di-tert-butyl-1-hydroxybenzene stearyl ester, ADK Stab AO 50, AO 4, AO 50, AT 76, Addiox 1016, Anox PP 18, Antioxidant 1076, Arenox A 76, Chemnox 1076, Chinox 1076, E 376, Hostanox 016, Hostanox M 016, Hostanox O 16, I 1076, IR 1076, IX 1076, Irganox 1068, Irganox 1076, Irganox 1076DWJ, Irganox 1906, Irganox 1976, Irganox I 1076, Irganox L 107, Irgastab PVC 76, JY 1076, KY 1076, L 107, M 4, Mark AO 50, Naugard 76, O 16, Octadecanyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl 3,5-bis(1,1-dimethylethyl)-4-hydroxyphenylpropanoate, Octadecyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, Octadecyl 3,5-di-tert-butyl-4-hydroxyphenylpropionate, Octadecyl 3,5-di-tert-butyl-4-oxyphenylpropionate, Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl 3-(3',5'-di-tert-butyl-4-hydroxyphenyl) propionate, Octadecyl 3-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Octadecyl 3-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, Octadecyl 3-[3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzene]propionate, Octadecyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl β-(3,5-tert-butyl-4-hydroxyphenyl)propionate, Octadecyl β-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Octadecyl β-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propenoate, Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate, Quantox 1076, Ralox 530, Selosol F 773, Songnox 1076, Sonox 1076, Stearyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, Stearyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Stearyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Sumilizer BP 76, Tinogard TS, Tominokusu SS, Tominox SS, U 276, Ultranox 276, n-Octadecyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, n-Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, n-Octadecyl 3-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, n-Octadecyl β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, n-Octadecyl β-(4'-hydroxy-3',5'-di-tert-butylphenyl)propionate, n-Octadecyl-3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate
IUPAC-namn octadecyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
LEDER CCCCCCCCCCCCCCCCCCOC(=O)CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
Specifikationer
Kemiskt namn eller material Plastic additive 11
Kvantitet 50 g
InChI formel InChI=1S/C35H62O3/c1-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-26-38-32(36)25-24-29-27-30(34(2,3)4)33(37)31(28-29)35(5,6)7/h27-28,37H,8-26H2,1-7H3
Formel vikt 530.4699