missing translation for 'onlineSavingsMsg'
Läs mer

Oktadecyltrietoxysilan, TRC

Product Code. 30410832
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30410832 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410832 Supplier Toronto Research Chemicals Supplier No. TRCO1484931G
 Begränsad artikel
Beräknad leverans: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Octadecyltriethoxysilane
CAS 7399-00-0
Molekylformel C24 H52 O3 Si
Kvantitet 1 g
Synonym Triethoxyoctadecylsilane, LS 6970, ODES, P 09775, S 12325, SIO 6645.0, Si 118, Stearyltriethoxysilane, n-Octadecyltriethoxysilane
InChI formel InChI=1S/C24H52O3Si/c1-5-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-28(25-6-2,26-7-3)27-8-4/h5-24H2,1-4H3
LEDER CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
IUPAC-namn trietoxi(oktadecyl)silan
Molekylvikt (g/mol) 416.753
Formel vikt 416.369
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.