Läs mer
Octafluoroadiponitrile, TRC
Produktkod. 30421431
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421431 500 mg
30411805 50 mg
30414041 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421431 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO273095500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 376-53-4
Molekylformel C6N2F8
Molekylvikt (g/mol) 252.07
IUPAC-namn 2,2,3,3,4,4,5,5-octafluorohexanedinitrile
LEDER FC(F)(C#N)C(F)(F)C(F)(F)C(F)(F)C#N
Specifikationer
Kemiskt namn eller material Octafluoroadiponitrile
Kvantitet 500 mg
InChI formel InChI=1S/C6F8N2/c7-3(8,1-15)5(11,12)6(13,14)4(9,10)2-16
Formel vikt 251.9934