Läs mer
Octafluorocyclopentene, TRC
Produktkod. 30423119
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30423119 500 mg
1 options
Produktkod. 30423119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO273120500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Octafluorocyclopentene
CAS 559-40-0
Molekylformel C5F8
Kvantitet 500 mg
Synonym Cyclopentene','1,2,3,3,4,4,5,5-octafluoro- (ACI), 1,2,3,3,4,4,5,5-Octafluorocyclopentene (ACI), Cyclopentene','octafluoro- (6CI','7CI','8CI','9CI), 1,2,3,3,4,4,5,5-Octafluoro-1-cyclopentene, Flon C 1418, Fron C 1418, Octafluorocyclopentene, Perfluorocyclopentene, Zeorora ZFL 58
InChI formel InChI=1S/C5F8/c6-1-2(7)4(10,11)5(12,13)3(1,8)9
LEDER FC1=C(F)C(F)(F)C(F)(F)C1(F)F
IUPAC-namn 1,2,3,3,4,4,5,5-octafluorocyclopentene
Molekylvikt (g/mol) 212.04
Formel vikt 211.9872