missing translation for 'onlineSavingsMsg'
Learn More

Octafluorotoluene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081431
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
434-64-0
C7F8
236.06
MFCD00000375
USPWUOFNOTUBAD-UHFFFAOYSA-N
octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
9906
1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
This item is not returnable. View return policy
434-64-0
C7F8
236.06
MFCD00000375
USPWUOFNOTUBAD-UHFFFAOYSA-N
octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
9906
1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -66°C
Density 1.668
Boiling Point 103°C to 105°C
Flash Point 20°C (68°F)
Refractive Index 1.368
Quantity 5 g
UN Number UN1992
Beilstein 2057072
Formula Weight 236.06
Percent Purity ≥98%
Chemical Name or Material Octafluorotoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'