Learn More
Lenz Laborglasintrumente™ Octagonal Plate Stoppers
Octagonal stopper made from solid DURAN™ glass
Brand: Lenz Laborglasintrumente™ 01400510
Additional Details : Weight : 0.80000kg
Description
- DIN 12252
- Solid glass
- Pack of 10
Specifications
Solid | |
10 | |
Borosilicate Glass | |
NS 10/19 |