Learn More
Fisherbrand™ Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
99.02 SEK - 391.00 SEK
Product Code | Brand | Length (Metric) | Diameter (Metric) | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Length (Metric) | Diameter (Metric) | Precio | Cantidad y disponibilidad | ||||
11577772
|
Fisherbrand™
|
13 mm | 8 mm |
99.02 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
11507782
|
Fisherbrand™
|
38 mm | 8 mm |
225.54 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
11537782
|
Fisherbrand™
|
38 mm | 9.5 mm |
240.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
11547782
|
Fisherbrand™
|
51 mm | 9.5 mm |
314.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
11527782
|
Fisherbrand™
|
25 mm | 9.5 mm |
196.66 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
11557782
|
Fisherbrand™
|
64 mm | 9.5 mm |
391.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
Descripción
- Stable, general-purpose magnetic stir bar
- The flat surfaces provide a greater mixing action
- All have pivot rings
- AlNiCo-V magnets encased in PTFE
- Autoclavable
Especificaciones
10/Pk. |