Läs mer
Octamethyl-1,7-tetrasiloxanediol, TRC
Produktkod. 30411249
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411249 250 mg
30412134 25 mg
30414331 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411249 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO860130250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3081-07-0
Molekylformel C8H26O5Si4
Molekylvikt (g/mol) 314.63
Synonym 1,1,3,3,5,5,7,7-Octamethyl-1,7-tetrasiloxanediol, 1,7-Dihydroxyoctamethyltetrasiloxane
IUPAC-namn hydroxy-[[[hydroxy(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane
LEDER C[Si](C)(O)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O
Specifikationer
Kemiskt namn eller material Octamethyl-1,7-tetrasiloxanediol
Kvantitet 250 mg
InChI formel InChI=1S/C8H26O5Si4/c1-14(2,9)11-16(5,6)13-17(7,8)12-15(3,4)10/h9-10H,1-8H3
Formel vikt 314.0857