missing translation for 'onlineSavingsMsg'
Learn More

Octan-2-ol, 98+%, Pure, Fisher Chemical™

Catalog Number p-7073349
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy
123-96-6
C8H18O
130.231
4591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O
This item is not returnable. View return policy
123-96-6
C8H18O
130.231
4591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O

C8H18O, CAS Number-123-96-6, sec-caprylic alcohol, n-octan-2-ol, methylhexylcarbinol, hexylmethylcarbinol, 2-octyl alcohol, 2-hydroxyoctane, dl-2-octanol, 2-octanol, 1-methylheptyl alcohol, 1-methylheptanol, 2.5L, 174 deg.C, CHEBI:37869, Yellow, 130.23g/mol, SJWFXCIHNDVPSH-UHFFFAOYSA-N, -38 deg.C

TRUSTED_SUSTAINABILITY
Melting Point -38°C
Color Yellow
Boiling Point 174°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 130.23g/mol
Physical Form Liquid
compliance-icons
Product Identifier
  • DL-2-Octanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'