Läs mer
Octanoyl L-Carnitine Chloride, TRC
Produktkod. 30423447
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423447 1 g
30410588 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423447 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO2380051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54377-02-5
Molekylformel C15H30ClNO4
Molekylvikt (g/mol) 323.86
Synonym (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctyl)oxy]-1-propanaminium Chloride
IUPAC-namn [(2R)-3-carboxy-2-octanoyloxypropyl]-trimethylazanium;chloride
LEDER O=C(O[C@@H](C[N+](C)(C)C)CC(O)=O)CCCCCCC.[Cl-]
Specifikationer
Kemiskt namn eller material Octanoyl L-Carnitine Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C15H29NO4.ClH/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4;/h13H,5-12H2,1-4H3;1H/t13-;/m1./s1
Formel vikt 323.1863
Procent renhet >95%