Läs mer
(±)-Octanoylcarnitine Chloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (±)-Octanoylcarnitine Chloride |
| CAS | 18822-86-1 |
| Molekylformel | C15H30NO4+ • Cl- |
| Kvantitet | 5 mg |
| Synonym | (3-Carboxy-2-hydroxypropyl)trimethylammonium Chloride','Octanoate (7CI), 3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctyl)oxy]-1-propanaminium','Chloride (9CI), (3-carboxy-2-hydroxypropyl)trimethylammonium','Chloride','Octanoate (8CI), Octanoic Acid','Ester with (3-Carboxy-2-hydroxypropyl)trimethylammonium Chloride, Octanoyl Carnitine Hydrochloride, Octanoyl-dl-carnitine Hydrochloride, Octanoylcarnitine Chloride |
| InChI formel | InChI=1S/C15H29NO4.ClH/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4;/h13H,5-12H2,1-4H3;1H |
| LEDER | O=C(OC(C[N+](C)(C)C)CC(O)=O)CCCCCCC.[Cl-] |
| Molekylvikt (g/mol) | 288.403545 |