Läs mer
MedChemExpress Octreotide
Produktkod. 16412619
Encompass_Preferred
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
16412619 1 mg
1 options
Produktkod. 16412619 Leverantör MedChemExpress Leverantörsnummer HYP0036
MedChemExpress Octreotide (SMS 201-995) is a somatostatin receptor agonist and synthetic octapeptide endogenous somatostatin analogue. Octreotide (SMS 201-995) can bind to the somatostatin receptor and mainly subtypes 2, 3, and 5, increases Gi activity, and reduces intracellular cAMP production. Octreotide (SMS 201-995) has antitumor activity, mediates apoptosis and may also be used in disease studies in acromegaly.
Specifikationer
Renhet 98.84%
CAS 83150-76-9
Innehåll och lagring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣The compound is unstable in solutions, freshly prepared is recommended.
För användning med (applikation) COVID-19-immunoregulation
Molekylvikt (g/mol) 1019.24 g/mol
Produkttyp Peptides
Kvantitet 1 mg
Frakt skick Room temperature
Löslighet H2O : 100 mg/mL (98.11 mM∣Need ultrasonic)
Synonym SMS 201-995