Läs mer
Octyl Decyldimethyl Ammonium Chloride, TRC
Produktkod. 17902715
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17902715 1 g
17972255 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17902715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO2399701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32426-11-2
Molekylformel C20 H44 N . Cl
Molekylvikt (g/mol) 334.02
Synonym 1-Decanaminium, N,N-dimethyl-N-octyl-, chloride (9CI), Ammonium, decyldimethyloctyl-, chloride (8CI), BTC 818, Bardac 2050, Decyldimethyloctylammonium chloride, Decyloctyldimethylammonium chloride, N-Octyl-N-decyl-N-dimethyl-ammonium chloride, Octyldecyldimethylammonium chloride, Quaternium 24
IUPAC-namn decyl-dimethyl-octylazanium;chloride
LEDER [Cl-].CCCCCCCCCC[N+](C)(C)CCCCCCCC
Specifikationer
Kemiskt namn eller material Octyl Decyldimethyl Ammonium Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C20H44N.ClH/c1-5-7-9-11-13-14-16-18-20-21(3,4)19-17-15-12-10-8-6-2;/h5-20H2,1-4H3;1H/q+1;/p-1
Formel vikt 333.3162
Procent renhet >95%