missing translation for 'onlineSavingsMsg'
Learn More

Octyl Decyldimethyl Ammonium Chloride, TRC

Product Code. 17972255
Change view
Click to view available options
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17972255 100 mg
17902715 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17972255 Supplier Toronto Research Chemicals Supplier No. TRCO239970100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32426-11-2
Molekylformel C20 H44 N . Cl
Molekylvikt (g/mol) 334.02
Synonym 1-Decanaminium, N,N-dimethyl-N-octyl-, chloride (9CI), Ammonium, decyldimethyloctyl-, chloride (8CI), BTC 818, Bardac 2050, Decyldimethyloctylammonium chloride, Decyloctyldimethylammonium chloride, N-Octyl-N-decyl-N-dimethyl-ammonium chloride, Octyldecyldimethylammonium chloride, Quaternium 24
IUPAC-namn decyl-dimethyl-octylazanium;chloride
LEDER [Cl-].CCCCCCCCCC[N+](C)(C)CCCCCCCC

Specifications

Kemiskt namn eller material Octyl Decyldimethyl Ammonium Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H44N.ClH/c1-5-7-9-11-13-14-16-18-20-21(3,4)19-17-15-12-10-8-6-2;/h5-20H2,1-4H3;1H/q+1;/p-1
Formel vikt 333.3162
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.