Läs mer
Octyl Dimethyl PABA, TRC
Produktkod. 30414300
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30414300 100 mg
1 options
Produktkod. 30414300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO241160100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Octyl Dimethyl PABA
CAS 58817-05-3
Anteckningar om renhetsgrad HPLC
Molekylformel C17H27NO2
Kvantitet 100 mg
Synonym p-(Dimethylamino)benzoic Acid Octyl Ester, Amerscol, Octyl N,N-Dimethyl-p-aminobenzoate, Octyl Dimethyl-p-aminobenzoate, Octyl p-Dimethylaminobenzoate
InChI formel InChI=1S/C17H27NO2/c1-4-5-6-7-8-9-14-20-17(19)15-10-12-16(13-11-15)18(2)3/h10-13H,4-9,14H2,1-3H3
LEDER O=C(OCCCCCCCC)C1=CC=C(N(C)C)C=C1
IUPAC-namn octyl 4-(dimethylamino)benzoate
Molekylvikt (g/mol) 277.4
Formel vikt 277.2042