missing translation for 'onlineSavingsMsg'
Learn More

ODAM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen ODAM
Concentration 0.37 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16836164
View Documents Promotion Details
Proteintech
16509-1-AP-20UL
20 μL
1656.73 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16826164
View Documents Promotion Details
Proteintech
16509-1-AP-150UL
150 μL
4721.67 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
Specifications

Specifications

ODAM
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
A1E959
54959
ODAM Fusion Protein Ag9726
Primary
-20°C
ODAM
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
APIN, ODAM
ODAM
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications