missing translation for 'onlineSavingsMsg'
Learn More

ODAM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16509-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826164

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
Specifications

Specifications

ODAM
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1E959
APIN, ODAM
Rabbit
IgG
150 μL
-20°C
Primary
54959
Immunohistochemistry (Paraffin)
Unconjugated
ODAM
Liquid
ODAM
ODAM Fusion Protein Ag9726
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers