missing translation for 'onlineSavingsMsg'
Learn More

OGN Mouse anti-Human, Porcine, Clone: 2A5F11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66382-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888205

  • 3313.85 SEK / 150µL
Estimated Shipment: 31-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.
Specifications

Specifications

OGN
2A5F11
Unconjugated
OGN
Liquid
OGN
OGN Fusion Protein Ag3484
Protein G
RUO
Antibody
Monoclonal
Human, Porcine
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20774
Mimecan, OG, OGN, OIF, osteoglycin, Osteoinductive factor, SLRR3A
Mouse
IgG1
150 μL
-20°C
Primary
106509723, 4969
Videos
SDS
Documents
Certificates
Special Offers

Special Offers