Learn More
OHAUS™ CS Series Compact Scales
Lightweight, portable electronic scales, ideal for work outside of the lab
657.40 SEK - 742.83 SEK
Specifications
Type CS Compact Scale
Stabilization Time 3 sec.
View More Specs
Includes: Three AA batteries, Stainless-steel weighing pan, AC adapter
Products 3
Product Code Brand Capacity Includes Model Readability Price Quantity & Availability  
Product Code Brand Capacity Includes Model Readability Price Quantity & Availability  
11848682 View accessory products OHAUS™
83997618
2 kg (72 oz.) Postal charts, AC adapter, three AA batteries, stainless-steel weighing pan CS2000P 1.0g 657.40 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
12-07-2019
Add to basket
11838682 View accessory products OHAUS™
83997617
200 g AC adapter, three AA batteries, stainless-steel weighing pan CS200-001 0.1g 657.40 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
04-07-2019
Add to basket
10773007 View accessory products OHAUS™
83997619
5 kg, 11.02 lb. AC adapter, three AA batteries, stainless-steel weighing pan CS5000-001 1g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
Suited for a wide range of applications, including quality control, formulations, postal weighing, forensic testing and soil and water sampling, and dietary needs.
  • Easy to operate with simple two-button operation with standard stainless steel weighing pan
  • Three low-profile models with weighing capacities from 200 to 5000g Low-battery indicator, overload/underload indicators, tactile feedback keyboard and auto shutoff Quick, easy calibration allows user to adjust for changes in location and temperature
  • Rugged construction provides overload protection up to 150% of capacity Powered by three AA batteries or AC adapter (both included)
  • Weighs in g, lb:oz, Newton, oz
  • Net weight: 0.91 lb. (0.47kg)
  • Dimensions (L × W × H): 7.625 × 5.312 × 1.5 in. (19.3 × 13.5 × 3.9cm)
  • Manufactured in ISO 9001 registered facility
Service Plan
Service agreement available.
Certifications
ISO 9001 compliant
Warranty
One-year (U.S. and Canada)
Specifications
Specifications
CS Compact Scale
3 sec.