Learn More
OHAUS™ Impact Printer Paper
For SF-42 printer.
76.70 SEK - 116.73 SEK
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11855810 OHAUS™
12101511
Printer Paper 76.70 SEK
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment
08-05-2019
Add to basket
12777246 OHAUS™
12101512
Ribbon 116.73 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
08-05-2019
Add to basket
Description
Description
  • Width: 2.25 in. (5.7cm)
  • Quantity: 5/Pk.