Läs mer
Kern Sohn OIML Class E1 Weight Set, Wooden Box
Beskrivning
- Set of weights
- Polished stainless steel
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifikationer
Specifikationer
| Klassificering | E1 |
| Material | Stainless Steel |
| Material för fodral | Wood |
| Vikt (metrisk) | 1 mg to 100 g |