Läs mer
Kern Sohn OIML Class E2 Weight Safety Set
Produktkod. 17371953
Encompass_Preferred
Change view
Vikt (metrisk):
10, 100 g
10, 200 g
100 mg, 2 g
100, 2000 g
20, 200 g
20, 500 g
200 mg, 5 g
5, 100 g
5, 50 g
Klassificering:
E2
M1, E2
Förpackningsstorlek:
1 uppsättning
Varje
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Vikt (metrisk) Klassificering unitSize
17371953 20, 500 g E2 -
17381943 100 mg, 2 g E2 -
17391943 200 mg, 5 g E2 -
17301953 5, 50 g E2 1 uppsättning
17311953 5, 100 g E2 -
17321953 10, 100 g E2 1 uppsättning
17331953 10, 200 g E2 1 uppsättning
17341953 20, 200 g E2 -
17321963 100, 2000 g M1, E2 Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Produktkod. 17371953 Leverantör Kern Sohn Leverantörsnummer 315109105
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Safety set contains one test weight for testing the upper end of the weighing range of your balance and one test weight for testing the lower end.
  • These KERN Safety Sets each contain one test weight for testing the sensitivity, i.e. the correct adjustment of your balance as well as a small test weight to test the lower end of the weighing range, the so-called minimum sample weight
  • As an option, the KERN Safety Set has space for another test weight, for testing your balance at a weight which is relevant for you
  • Useful accessories which have been selected to suit that particular KERN Safety Set, such as, for example, special gloves, tweezers, weight grips, brushes, etc. will assist you in handling your test weights properly
  • Stored in the practical protective case next to your balance, you can check and ensure the high precision of your balance at any time
Specifikationer
Klassificering E2
Material Stainless Steel
Vikt (metrisk) 20, 500 g
Inkluderar Protective Case