Läs mer
Kern Sohn OIML Class F1 Weight Set, Wooden Box
Description
- Knob shape
- Polished stainless steel
- Lined case
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifications
Specifications
| Klassificering | F1 |
| Material | Stainless Steel |
| Material för fodral | Wood |
| Vikt (metrisk) | 1 mg to 5 kg |