Läs mer
Kern Sohn OIML Class F1 Weight Set, Wooden Box
Produktkod. 17392063
Encompass_Preferred
Change view
Vikt (metrisk):
1 g to 1 kg
1 g to 10 kg
1 g to 2 kg
1 g to 5 kg
1 mg to 1 kg
1 mg to 10 kg
1 mg to 100 g
1 mg to 2 kg
1 mg to 200 g
1 mg to 5 kg
1 mg to 50 g
1 mg to 500 g
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
Förpackningsstorlek:
1 uppsättning
Varje
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Vikt (metrisk) unitSize
17392063 1 g to 10 kg -
17302043 1 mg to 50 g -
17322043 1 mg to 100 g -
17342043 1 mg to 200 g Varje
17362043 1 mg to 500 g -
17382043 1 mg to 1 kg -
17302053 1 mg to 2 kg -
17322053 1 mg to 5 kg -
17342053 1 mg to 10 kg -
17352053 1 to 50 g 1 uppsättning
17372053 1 to 100 g 1 uppsättning
17392053 1 to 200 g -
17312063 1 to 500 g -
17332063 1 g to 1 kg -
17352063 1 g to 2 kg -
17372063 1 g to 5 kg 1 uppsättning
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Produktkod. 17392063 Leverantör Kern Sohn Leverantörsnummer 32409
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Weight set constructed of polished stainless steel and housed in a wooden box.
  • Knob shape
  • Polished stainless steel
  • Lined case
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifikationer
Klassificering F1
Material Stainless Steel
Material för fodral Wood
Vikt (metrisk) 1 g to 10 kg