Learn More
Ohaus™ OIML E2 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E2 Tolerances
8415.00 SEK - 57635.00 SEK
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16622452
|
Ohaus™
80780381 |
500 mg to 1 mg |
8415.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16632452
|
Ohaus™
80780383 |
50 g to 1 mg |
18465.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16642452
|
Ohaus™
80780384 |
50 g to 1 g |
10855.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16652452
|
Ohaus™
80780385 |
100 g to 1 mg |
20355.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16662452
|
Ohaus™
80780386 |
100 g to 1 g |
12545.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16672452
|
Ohaus™
80780387 |
2 x 200 g to 1 mg |
22605.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16682452
|
Ohaus™
80780388 |
2 x 200 g to 1 g |
16805.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16692452
|
Ohaus™
80780391 |
500 g to 1 mg |
27100.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16602462
|
Ohaus™
80780392 |
500 g to 1 g |
20000.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16612462
|
Ohaus™
80780393 |
1 kg to 1 mg |
33375.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16622462
|
Ohaus™
80780394 |
1 kg to 1 g |
25565.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16632462
|
Ohaus™
80780395 |
2 x 2 kg to 1 mg |
47575.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16642462
|
Ohaus™
80780396 |
2 x 2 kg to 1 g |
40000.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16652462
|
Ohaus™
80780399 |
5 kg to 1 mg |
57635.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16662462
|
Ohaus™
80780400 |
5 kg to 1 g |
50060.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- Weight Sets in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |