Learn More
Ohaus™ OIML F2 Weight Set
Product Code. 16692202 Shop All Ohaus Products
Change view
Click to view available options
Mass:
1 kg to 1 g
1 kg to 1 mg
100 g to 1 g
100 g to 1 mg
2 x 2 kg to 1 g
2 x 2 kg to 1 mg
2 x 200 g to 1 g
2 x 200 g to 1 mg
5 kg to 1 g
5 kg to 1 mg
50 g to 1 g
50 g to 1 mg
500 g to 1 g
500 g to 1 mg
500 mg to 1 mg
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass Unit Size
16692202 100 g to 1 mg Each
16622212 500 mg to 1 mg Each
16612212 50 g to 1 mg Each
16602212 50 g to 1 g Each
16682202 100 g to 1 g Each
16632192 2 x 200 g to 1 mg Each
16622192 2 x 200 g to 1 g Each
16672202 500 g to 1 mg Each
16662202 500 g to 1 g Each
16652202 1 kg to 1 mg Each
16642202 1 kg to 1 g Each
16612192 2 x 2 kg to 1 mg Each
16602192 2 x 2 kg to 1 g Each
16632202 5 kg to 1 mg Each
16622202 5 kg to 1 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 16692202 Supplier Ohaus™ Supplier No. 30390825
  • ONLINE EXCLUSIVE PRICE
    7870.00 SEK
    5454.81 SEK / Paket med 1
    Save 2415.19 SEK
    30% Av
Beräknad leverans: 17-12-2026
Adjusted to Meet or Exceed OIML F2 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance No Certificate
Material Stainless Steel, Aluminum
Mass 100 g to 1 mg
Includes Protective Case
Packaging Quantity 21/Pk.