Learn More
Ohaus™ OIML F2 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F2 Tolerances
3945.00 SEK - 24735.00 SEK
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16632222
|
Ohaus™
30390852 |
500 mg to 1 mg |
3945.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16622222
|
Ohaus™
30390850 |
50 g to 1 mg |
8725.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16612222
|
Ohaus™
30390849 |
50 g to 1 g |
4500.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16602222
|
Ohaus™
30390848 |
100 g to 1 mg |
9575.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16692212
|
Ohaus™
30390847 |
100 g to 1 g |
5555.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16672192
|
Ohaus™
30390036 |
2 x 200 g to 1 mg |
11280.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16662192
|
Ohaus™
30390035 |
2 x 200 g to 1 g |
7505.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16682212
|
Ohaus™
30390844 |
500 g to 1 mg |
12665.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16672212
|
Ohaus™
30390843 |
500 g to 1 g |
8415.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16662212
|
Ohaus™
30390842 |
1 kg to 1 mg |
15505.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16652212
|
Ohaus™
30390841 |
1 kg to 1 g |
12430.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16652192
|
Ohaus™
30390034 |
2 x 2 kg to 1 mg |
20710.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16642192
|
Ohaus™
30390033 |
2 x 2 kg to 1 g |
20595.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16642212
|
Ohaus™
30390838 |
5 kg to 1 mg |
24735.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
16632212
|
Ohaus™
30390837 |
5 kg to 1 g |
20830.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |