missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OIP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12142-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
OIP5 | |
Polyclonal | |
Unconjugated | |
OIP5 | |
5730547N13Rik, Cancer/testis antigen 86, CT86, LINT 25, MIS18B, MIS18beta, OIP 5, OIP5, Opa interacting protein 5, Protein Mis18 beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11339 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43482 | |
OIP5 | |
OIP5 Fusion Protein Ag2790 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |