missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Olanzapine EP Impurity C (as Chloride), Mikromol™
Beskrivning
Olanzapine EP Impurity C (as Chloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium Chloride (Olanzapine-N-Chloromethyl Chloride) |
| CAS | 719300-59-1 |
| Analyt- eller komponentnamn | 1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium Chloride (Olanzapine-N-Chloromethyl Chloride) |
| Frakt skick | Room Temperature |
| Molekylformel | C18 H22 Cl N4 S . Cl |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Olanzapine Imp. C (EP), 1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium Chloride, Olanzapine-N-Chloromethyl Chloride, Olanzapine Impurity C as Chloride |
| InChI formel | InChI=1S/C18H22ClN4S.ClH/c1-13-11-14-17(22-7-9-23(2,12-19)10-8-22)20-15-5-3-4-6-16(15)21-18(14)24-13;/h3-6,11,21H,7-10,12H2,1-2H3;1H/q+1;/p-1 |
| LEDER | [Cl-].Cc1cc2C(=Nc3ccccc3Nc2s1)N4CC[N+](C)(CCl)CC4 |
| IUPAC-namn | 4-[4-(chloromethyl)-4-methylpiperazin-4-ium-1-yl]-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine;chloride |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?