Läs mer
Olcegepant hydrochloride, MedChemExpress
Produktkod. 16435957
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16435957 5 mg
16423228 10 mg
16419776 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16435957 Leverantör Medchem Express Leverantörsnummer HY10095A
MedChemExpress Olcegepant hydrochloride (BIBN-4096 hydrochloride) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and with a Ki of 14.4 pM for human CGRP.
Olcegepant hydrochloride (BIBN-4096 hydrochloride) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and with a Ki of 14.4 pM for human CGRP.
Specifikationer
Kemiskt namn eller material Olcegepant hydrochloride
CAS 586368-06-1
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C38H47Br2N9O5.HCl
Kvantitet 5 mg
Synonym BIBN-4096 hydrochloride BIBN4096BS hydrochloride
Löslighetsinformation DMSO : 100 mg/mL (110.36 mM; Need ultrasonic) ∣H2O : ≥ 66.66 mg/mL (73.57 mM)
LEDER O=C1N(CC2=C(N1)C=CC=C2)C3CCN(C(N[C@@H](C(N[C@H](C(N4CCN(C5=CC=NC=C5)CC4)=O)CCCCN)=O)CC6=CC(Br)=C(C(Br)=C6)O)=O)CC3.Cl
Molekylvikt (g/mol) 906.11
Formel vikt 906.11
Procent renhet 99.11%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Neuroscience-Neuromodulation