Läs mer
Oleandrin, TRC
Produktkod. 30040701
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040701 10 mg
30040481 1 mg
30040433 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040701 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO25800010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 465-16-7
Molekylformel C32 H48 O9
Molekylvikt (g/mol) 576.72
Synonym Oleandrin (6CI,7CI,8CI), (3β,5β,16β)-16-(Acetyloxy)-3-[(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)oxy]-14-hydroxycard-20(22)-enolide, Corrigen, Foliandrin, Folinerin, Neriol, Neriolin, Neriostene, Oleandrigenin 3-O-α-L-oleandroside, Oleandrine
IUPAC-namn [(3S,8R,9S,10S,13R,14S,16S,17R)-14-hydroxy-3-[(2R,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-16-yl] acetate
LEDER CO[C@H]1C[C@H](O[C@H]2CC[C@@]3(C)C(CC[C@@H]4[C@@H]3CC[C@]5(C)[C@H]([C@H](C[C@]45O)OC(=O)C)C6=CC(=O)OC6)C2)O[C@@H](C)[C@@H]1O
Specifikationer
Kemiskt namn eller material Folinerin (Oleandrin)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C32H48O9/c1-17-29(35)24(37-5)14-27(39-17)41-21-8-10-30(3)20(13-21)6-7-23-22(30)9-11-31(4)28(19-12-26(34)38-16-19)25(40-18(2)33)15-32(23,31)36/h12,17,20-25,27-29,35-36H,6-11,13-16H2,1-5H3/t17-,20?,21-,22-,23+,24-,25-,27-,28-,29-,30-,31+,32-/m0/s1
Formel vikt 576.3298
Procent renhet >95%