Läs mer
Oleanonic acid, MedChemExpress
Produktkod. 16474917
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16474917 50 mg
1 options
Produktkod. 16474917 Leverantör Medchem Express Leverantörsnummer HYN1487
MedChemExpress Oleanonic acid (3-Oxooleanolic acid) is a triterpenoid, inhibits infection by HIV-1 in in vitro infected PBMC, naturally infected PBMC and monocyte/macrophages with EC50 of 22.7 mM, 24.6 mM and 57.4 mM, respectively.
Oleanonic acid (3-Oxooleanolic acid) is a triterpenoid, inhibits infection by HIV-1 in in vitro infected PBMC, naturally infected PBMC and monocyte/macrophages with EC50 of 22.7 mM, 24.6 mM and 57.4 mM, respectively.
Specifikationer
Kemiskt namn eller material Oleanonic acid
Fysisk form Solid
CAS 17990-42-0
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C30H46O3
För användning med (applikation) Cancer-programmed cell death
Synonym 3-Oxooleanolic acid
Löslighetsinformation DMSO : 50 mg/mL (109.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER CC1(C)C(CC[C@]2(C)[C@@]3([H])CC=C4[C@]5([H])CC(C)(C)CC[C@@](C(O)=O)5CC[C@](C)4[C@@](C)3CC[C@@]12[H])=O
Molekylvikt (g/mol) 454.68
Formel vikt 454.68
Procent renhet 98.0%