Läs mer
Oleanonic Acid, TRC
Produktkod. 30411348
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30411348 500 mg
1 options
Produktkod. 30411348 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO521005500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oleanonic Acid
CAS 17990-42-0
Molekylformel C30 H46 O3
Kvantitet 500 mg
Synonym 3-Oxoolean-12-en-28-oic Acid, 3-Ketooleanolic Acid, 3-Oxooleanolic acid, BLE 99006, STX 50
InChI formel InChI=1S/C30H46O3/c1-25(2)14-16-30(24(32)33)17-15-28(6)19(20(30)18-25)8-9-22-27(5)12-11-23(31)26(3,4)21(27)10-13-29(22,28)7/h8,20-22H,9-18H2,1-7H3,(H,32,33)/t20-,21-,22+,27-,28+,29+,30-/m0/s1
LEDER CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CCC(=O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O
IUPAC-namn (4aS,6aR,6aS,6bR,8aR,12aR,14bS)-2,2,6a,6b,9,9,12a-heptamethyl-10-oxo-3,4,5,6,6a,7,8,8a,11,12,13,14b-dodecahydro-1H-picene-4a-carboxylic acid
Molekylvikt (g/mol) 454.68
Formel vikt 454.3447