Läs mer
Oleic Acid N-Hydroxysuccinimide, TRC
Produktkod. 17921715
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17921715 1 g
17915015 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17921715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO5243001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81480-40-2
Molekylformel C22 H37 N O4
Molekylvikt (g/mol) 379.53
Synonym (9Z)-9-Octadecenoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, (Z)-1-[(1-Oxo-9-octadecenyl)oxy]-2,5-pyrrolidinedione, 1-[[(9Z)-1-Oxo-9-octadecenyl]oxy]-2,5-pyrrolidinedione, N-Hydroxysuccinimide Ester with Oleic Acid, N-Hydroxysuccinimide Oleate, N-Succinimidyl Oleate, Oleic Acid N-Hydroxysuccinimide Ester, Succinimidyl Oleate
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) (Z)-octadec-9-enoate
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)ON1C(=O)CCC1=O
Specifikationer
Kemiskt namn eller material Oleic Acid N-Hydroxysuccinimide
Kvantitet 1 g
InChI formel InChI=1S/C22H37NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(26)27-23-20(24)18-19-21(23)25/h9-10H,2-8,11-19H2,1H3/b10-9-
Formel vikt 379.2723