Läs mer
Oleoyl Chloride, TRC
Produktkod. 30423437
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30423437 500 mg
1 options
Produktkod. 30423437 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO526750500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oleoyl Chloride
CAS 112-77-6
Molekylformel C18 H33 Cl O
Kvantitet 500 mg
Synonym Oleoyl Chloride, (Z)-9-Octadecenoyl Chloride, 9-(Z)-Octadecen-1-oyl Chloride, Oleic Acid Chloride, Oleic Chloride, Oleyl Chloride, cis-9-Octadecenoyl Chloride
InChI formel InChI=1S/C18H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3/b10-9-
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)Cl
IUPAC-namn (Z)-octadec-9-enoyl chloride
Molekylvikt (g/mol) 300.907
Formel vikt 300.222