Läs mer
Oleuropein (>75%), TRC
Produktkod. 30009140
Change view
Kvantitet:
25 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009140 500 mg
30005058 10 g
30007654 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO532950500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32619-42-4
Molekylformel C12 H17 O7
Molekylvikt (g/mol) 540.51
Synonym (2S,3E,4S)-3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-Pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester, 2H-Pyran-4-acetic acid, 3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-, 2-(3,4-dihydroxyphenyl)ethyl ester, [2S-(2α,3E,4β)]-, 2H-Pyran-4-acetic acid, 5-carboxy-3-ethylidene-2-(β-D-glucosyloxy)-3,4-dihydro-, 3,4-dihydroxyphenethyl 5-methyl ester (7CI), Oleuropein (8CI), Oleoeuropein, Oleoeuropeine, Oleuropeine
IUPAC-namn methyl (4S,5E,6S)-4-[2-[2-(3,4-dihydroxyphenyl)ethoxy]-2-oxoethyl]-5-ethylidene-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4H-pyran-3-carboxylate
LEDER COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)\[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3
Specifikationer
Kemiskt namn eller material Oleuropein
Kvantitet 500 mg
InChI formel InChI=1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1
Formel vikt 540.1843