Läs mer
Oleuropein Aglycone, TRC
Produktkod. 30138314
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138314 50 mg
30133055 100 mg
30005396 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138314 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO53294550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31773-95-2
Molekylformel C19H22O8
Molekylvikt (g/mol) 378.37
Synonym (2R,3E,4S)-3-Ethylidene-3,4-dihydro-2-hydroxy-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester, [2R-(2α,3E,4β)]-3-Ethylidene-3,4-dihydro-2-hydroxy-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester, 5-Carboxy-3-ethylidene-3,4-dihydro-2-hydroxy-2H-pyran-4-acetic Acid 4-(3,4-Dihydroxyphenethyl) 5-Methyl Ester, 3,4-DHPEA-EA, Oleoeuropein Aglycone, Oleoeuropeine Aglycone, Oleuropein Aglycon, Oleuropeine Aglycone
IUPAC-namn methyl (4S,5E,6R)-4-[2-[2-(3,4-dihydroxyphenyl)ethoxy]-2-oxoethyl]-5-ethylidene-6-hydroxy-4H-pyran-3-carboxylate
LEDER O=C(OCCC1=CC=C(O)C(O)=C1)C[C@H]2/C([C@H](O)OC=C2C(OC)=O)=C\C
Specifikationer
Kemiskt namn eller material Oleuropein Aglycone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C19H22O8/c1-3-12-13(14(18(23)25-2)10-27-19(12)24)9-17(22)26-7-6-11-4-5-15(20)16(21)8-11/h3-5,8,10,13,19-21,24H,6-7,9H2,1-2H3/b12-3+/t13-,19+/m0/s1
Formel vikt 378.1315
Procent renhet >95%