Läs mer
Oleyl Acetate, TRC
Produktkod. 30420748
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30420748 2.5 g 2.5g
30417347 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420748 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO5329902.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oleyl Acetate
CAS 693-80-1
Molekylformel C20H38O2
Kvantitet 2.5 g
InChI formel InChI=1S/C20H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22-20(2)21/h10-11H,3-9,12-19H2,1-2H3/b11-10-
LEDER CCCCCCCC\C=C/CCCCCCCCOC(=O)C
IUPAC-namn [(Z)-octadec-9-enyl] acetate
Molekylvikt (g/mol) 310.51
Formel vikt 310.2872