missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Olig 1, 2, 3 Mouse anti-Human, Clone: 257224, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB2230-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Olig1/2/3 Monoclonal specifically detects Olig1/2/3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifikationer
| Olig1/2/3 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| BHLHB1, bHLHb6, BHLHB7, bHLHe21, PRKCBP2, RACK17 | |
| E. coli-derived recombinant human Olig1 aa 1-272 | |
| 25 μg | |
| Primary | |
| Detects human Olig 1, 2, 3. In direct ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Olig2 and 50 - 100% cross-reactivity with rhOlig3. | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry | |
| 257224 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 116448 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?