missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLIG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13999-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a basic helix-loop-helix transcription factor which is expressed in oligodendroglial tumors of the brain. The protein is an essential regulator of ventral neuroectodermal progenitor cell fate. The gene is involved in a chromosomal translocation t(14;21)(q11.2;q22) associated with T-cell acute lymphoblastic leukemia. Its chromosomal location is within a region of chromosome 21 which has been suggested to play a role in learning deficits associated with Down syndrome. Tissue specificity: Expressed in the brain, in oligodendrocytes. Strongly expressed in oligodendrogliomas, while expression is weak to moderate in astrocytomas. Expression in glioblastomas highly variable.Specifications
OLIG2 | |
Polyclonal | |
Unconjugated | |
OLIG2 | |
BHLHB1, bHLHe19, OLIG2, OLIGO2, PRKCBP2, RACK17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10215, 304103, 50913 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13516, Q9EQW6 | |
Olig2 | |
OLIG2 Fusion Protein Ag5089 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |