Läs mer
Olivetol Dimethyl Ether, TRC
Produktkod. 30408992
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30408992 100 mg
1 options
Produktkod. 30408992 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO533010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Olivetol Dimethyl Ether
CAS 22976-40-5
Molekylformel C13H20O2
Kvantitet 100 mg
Synonym 1,3-Dimethoxy-5-pentyl-benzene, 1,3-Dimethoxy-5-pentylbenzene, 3,5-Dimethoxyamylbenzene, 5-Pentylresorcinol Dimethyl Ether, Di-O-methylolivetol
InChI formel InChI=1S/C13H20O2/c1-4-5-6-7-11-8-12(14-2)10-13(9-11)15-3/h8-10H,4-7H2,1-3H3
LEDER CCCCCc1cc(OC)cc(OC)c1
IUPAC-namn 1,3-dimethoxy-5-pentylbenzene
Molekylvikt (g/mol) 208.3
Formel vikt 208.1463