Läs mer
Olivetol, TRC
Produktkod. 30424293
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424293 1 g
30423150 25 g
30409791 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424293 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO5330001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 500-66-3
Molekylformel C11 H16 O2
Molekylvikt (g/mol) 180.24
Synonym Olivetol, Olivetol (6CI), Resorcinol, 5-pentyl- (8CI), 5-Pentyl-1,3-benzenediol, 1,3-Dihydroxy-5-pentylbenzene, 3,5-Dihydroxy-1-pentylbenzene, 3,5-Dihydroxyamylbenzene, 5-Pentylbenzene-1,3-diol, 5-Pentylresorcinol, 5-n-Amylresorcinol, 5-n-Pentylresorcinol
IUPAC-namn 5-pentylbenzene-1,3-diol
LEDER CCCCCc1cc(O)cc(O)c1
Specifikationer
Kemiskt namn eller material 3,5-dihydroxy-1-pentylbenzene
Kvantitet 1 g
InChI formel InChI=1S/C11H16O2/c1-2-3-4-5-9-6-10(12)8-11(13)7-9/h6-8,12-13H,2-5H2,1H3
Formel vikt 180.115